missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD1B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SETD1B.
Brand: Abnova PAB21443.100uL
This item is not returnable.
View return policy
Description
SET1B is a component of a histone methyltransferase complex that produces trimethylated histone H3 at Lys4 (Lee et al., 2007 [PubMed 17355966]).[supplied by OMIM
Sequence: DSLGMEEEVDIETEAVAPEERPSMLDEPPLPVGVEEPADSREPPEEPGLSQEGAMLLSPEPPAKEVEARPPLSPERSpecifications
| SETD1B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SETD1B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ20803/KIAA1076/KMT2G/Set1B | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23067 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SETD1B | |
| SETD1B | |
| Recombinant protein corresponding to amino acids of human SETD1B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction