missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB14 (Ovalbumin), Rabbit, FITC, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Ovalbumin.
Brand: Abnova PAB19934.100ug
This item is not returnable.
View return policy
Specifications
| SERPINB14 (Ovalbumin) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Ovalbumin. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| SERPINB14 | |
| Ovalbumin [Hen Egg White] | |
| RUO | |
| 396058 | |
| Chicken | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence, Western Blot | |
| FITC | |
| ELISA (1:5000-1:20000) WB (1:2000-1:10000) IF (1:500-1:2500) The optimal working dilution should be determined by the end user. | |
| SERPINB14 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Ovalbumin [Hen Egg White]. Cross reactivity against Ovalbumin from other tissues and species may occur but have not been specifically determined. | |
| Store at 4°C on dry atmosphere. After reconstitution with 100μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction