Learn More
SERPINA3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SERPINA3.
Brand: Abnova PAB28589.100uL
Description
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this protein's sequence have been implicated in Alzheimer's disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease. [provided by RefSeq]
Sequence: QDKMEEVEAMLLPETLKRWRDSLEFREIGELYLPKFSISRDYNLNDILLQLGIEEAFTSKADLSGITGARNLAVSQVVHKAVLDVFEEGTEASAATAVKITLLSALVETRTIVRFNRPFLMIIVPTDTQNIFFMSKVTNPKQEGAPHRSpecifications
| SERPINA3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SERPINA3. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| AACT/ACT/GIG24/GIG25/MGC88254 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 12 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| SERPINA3 | |
| SERPINA3 | |
| Recombinant protein corresponding to amino acids of human SERPINA3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only