missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sepw1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant Sepw1.
Brand: Abnova PAB11324.100ug
This item is not returnable.
View return policy
Specifications
| Sepw1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant Sepw1. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| selW | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody is specific to mouse Selenoprotein W. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:100000) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
| Sepw1 | |
| Sepw1 | |
| Recombinant protein corresponding to full length mouse Sepw1. | |
| RUO | |
| 20364 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction