missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sept8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Sept8.
Brand: Abnova PAB15710.50ug
This item is not returnable.
View return policy
Description
Sequence: AVVGSTEEVKVGNKLVRARQYPWGVVQVENENHCDFVKLREMLIRVNMEDLREQTHSRHYELYRRCKLEEMGFQDSDGDSQPFSLQETYEAKRKEFLSELQRKEEEMRQMFVNKVKETELELKEKERELHEKFEHLKRIHQEEKRKVEEKRRELEEETNAFNCRKAAMEALQSQALHATSQQPLRKDKDKKNSpecifications
| Sept8. | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Sept8. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| AW046166/Sepl/mKIAA0202 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX0010. This antibody detects endogenous mSEPT8 protein in several cell types. It also recognizes human SEPT8 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Sept8 | |
| Sept8 | |
| Recombinant GST fusion protein corresponding to 192 mouse Sept8. | |
| RUO | |
| 20362 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction