Learn More
SENP1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SENP1.
Brand: Abnova PAB0343.100ug
Description
The covalent modification of proteins by the small ubiquitin (UBB; MIM 191339)-like protein SUMO (see SUMO1, MIM 601912) is implicated in the regulation of nucleocytoplasmic transport, genomic stability, gene transcription, and other processes. Sumoylation is catalyzed on target lysine residues by a multienzyme process and is reversed by desumoylating enzymes such as SENP1 (Yamaguchi et al., 2005 [PubMed 15923632]).[supplied by OMIM
Specifications
| SENP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SENP1. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| SuPr-2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| SENP1 | |
| SENP1 | |
| A synthetic peptide corresponding to amino acids 578-590 of human SENP1. | |
| RUO | |
| 29843 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only