missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sema5a, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Sema5a.
Brand: Abnova PAB2549.400uL
This item is not returnable.
View return policy
Specifications
| Sema5a | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Sema5a. | |
| In PBS (0.09% sodium azide) | |
| 5930434A13/9130201M22Rik/AI464145/Semaf/semF | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 20356 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Sema5a | |
| Sema5a | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of mouse Sema5a. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction