Learn More
SELENBP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SELENBP1.
Brand: Abnova PAB21102.100uL
Description
This gene product belongs to the selenium-binding protein family. Selenium is an essential nutrient that exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. It has been proposed that the effects of selenium in preventing cancer and neurologic diseases may be mediated by selenium-binding proteins. The exact function of this gene is not known. [provided by RefSeq
Sequence: IEPKDIHAKCELAFLHTSHCLASGEVMISSLGDVKGNGKGGFVLLDGETFEVKGTWERPGGAAPLGYDFWYQPRHNVMISTEWAAPNVLRDGFNPADVSpecifications
| SELENBP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SELENBP1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ13813/LPSB/SP56/hSBP/hSP56 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8991 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SELENBP1 | |
| SELENBP1 | |
| Recombinant protein corresponding to amino acids of human SELENBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only