Learn More
SEC31B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SEC31B.
Brand: Abnova PAB23494.100uL
Description
This gene encodes a protein of unknown function. The protein has moderate similarity to rat VAP1 protein which is an endosomal membrane-associated protein, containing a putative Ca2+/calmodulin-dependent kinase II phosphorylation site. [provided by RefSeq]
Sequence: HAQGSAVLGQQSPPFPFPRIVVGATLHSKETSSYRLGSQPSHQVPTPSPRPRVFTPQSSPAMPLAPSHPSPYQGPRTQNISDYRASpecifications
| SEC31B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SEC31B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp434M183/SEC31B-1/SEC31L2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 25956 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| SEC31B | |
| SEC31B | |
| Recombinant protein corresponding to amino acids of human SEC31B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only