Learn More
SDPR, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SDPR.
Brand: Abnova PAB27397.100ug
Description
This gene encodes a calcium-independent phospholipid-binding protein whose expression increases in serum-starved cells. This protein is a substrate for protein kinase C (PKC) phosphorylation and recruits polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation
Specifications
| SDPR | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| SDPR | |
| SDPR | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human SDPR. | |
| 100 μg | |
| Primary | |
| It is predicted to not cross-react with other members of the cavin family. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SDPR. | |
| In PBS (0.02% sodium azide) | |
| PS-p68/SDR | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 8436 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only