Learn More
SDC1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SDC1.
Brand: Abnova PAB20340.100uL
Description
The protein encoded by this gene is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan-1 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Altered syndecan-1 expression has been detected in several different tumor types. While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same protein. [provided by RefSeq
Sequence: TTTATTAQEPATSHPHRDMQPGHHETSTPAGPSQADLHTPHTEDGGPSATERAAEDGASSQLPAAEGSGEQDFTFETSGENTAVVAVEPDRRNQSPVDQSpecifications
| SDC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SDC1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CD138/SDC/SYND1/syndecan | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6382 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SDC1 | |
| SDC1 | |
| Recombinant protein corresponding to amino acids of human SDC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only