Learn More
SCYL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SCYL1.
Brand: Abnova PAB2778.400uL
Description
This gene encodes a transcriptional regulator belonging to the SCY1-like family of kinase-like proteins. The protein has a divergent N-terminal kinase domain that is thought to be catalytically inactive, and can bind specific DNA sequences through its C-terminal domain. It activates transcription of the telomerase reverse transcriptase and DNA polymerase beta genes. The protein has been localized to the nucleus, and also to the cytoplasm and centrosomes during mitosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| SCYL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SCYL1. | |
| In PBS (0.09% sodium azide) | |
| GKLP/HT019/MGC78454/NKTL/NTKL/P105/TAPK/TEIF/TRAP | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 57410 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SCYL1 | |
| SCYL1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SCYL1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only