Learn More
SCO2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SCO2.
Brand: Abnova PAB12984.100ug
Description
Mammalian cytochrome c oxidase (COX) catalyzes the transfer of reducing equivalents from cytochrome c to molecular oxygen and pumps protons across the inner mitochondrial membrane. In yeast, 2 related COX assembly genes, SCO1 and SCO2 (synthesis of cytochrome c oxidase), enable subunits 1 and 2 to be incorporated into the holoprotein. This gene is the human homolog of the yeast SCO2 gene. [provided by RefSeq
Specifications
| SCO2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SCO2. | |
| In PBS (0.02% sodium azide) | |
| MGC125823/MGC125825/SCO1L | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SCO2 | |
| SCO2 | |
| A synthetic peptide corresponding to C-terminus 19 amino acids of human SCO2. | |
| RUO | |
| 9997 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only