Learn More
SCNN1A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SCNN1A.
Brand: Abnova PAB20617.100uL
Description
Nonvoltage-gated, amiloride-sensitive, sodium channels control fluid and electrolyte transport across epithelia in many organs. These channels are heteromeric complexes consisting of 3 subunits: alpha, beta, and gamma. This gene encodes the alpha subunit, and mutations in this gene have been associated with pseudohypoaldosteronism type 1 (PHA1), a rare salt wasting disease resulting from target organ unresponsiveness to mineralocorticoids. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Sequence: RYPEIKEELEELDRITEQTLFDLYKYSSFTTLVAGSRSRRDLRGTLPHPLQRLRVPPPPHGARRARSVASSLRDNNPQVDWKDWKIGFQLSpecifications
| SCNN1A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SCNN1A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| ENaCa/ENaCalpha/FLJ21883/SCNEA/SCNN1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6337 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| SCNN1A | |
| SCNN1A | |
| Recombinant protein corresponding to amino acids of human SCNN1A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only