missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scnn1a, Rabbit, FITC, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.
Brand: Abnova PAB28778.100ug
This item is not returnable.
View return policy
Specifications
| Scnn1a | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Rat Scnn1a. | |
| PBS, 50% glycerol and 0.09% sodium azide | |
| Scnn1a | |
| Recombinant protein corresponding to Scnn1a N-terminus (amino acids 46-68). | |
| 100 μg | |
| Primary | |
| 1 ug/mL of PAB28778 was sufficient for detection of Scnn1a, ∼85kDa, in 35ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| FITC | |
| Immunofluorescence (1:400) Immunohistochemistry Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Scnn1a | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 25122 | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction