Learn More
SCN4B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SCN4B.
Brand: Abnova PAB20936.100uL
Description
The protein encoded by this gene is one of several sodium channel beta subunits. These subunits interact with voltage-gated alpha subunits to change sodium channel kinetics. The encoded transmembrane protein forms interchain disulfide bonds with SCN2A. Defects in this gene are a cause of long QT syndrome type 10 (LQT10). Three protein-coding and one noncoding transcript variant have been found for this gene
Sequence: LLPCTFSSCFGFEDLHFRWTYNSSDAFKILIEGTVKNEKSDPKVTLKDDDRITLVGSTKEKMNNISIVLRDLEFSDTGKYTCHVKNPKENNLQHHATIFLQVVSpecifications
| SCN4B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SCN4B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| SCN4B | |
| Recombinant protein corresponding to amino acids of human SCN4B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SCN4B | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6330 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only