missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCGB2A2, Rabbit, Clone: 31A5, Abnova™
Rabbit monoclonal antibody raised against full-length recombinant SCGB2A2.
Brand: Abnova MAB3550.500uL
This item is not returnable.
View return policy
Specifications
| SCGB2A2 | |
| Monoclonal | |
| 50 ug/mL | |
| Rabbit monoclonal antibody raised against full length recombinant SCGB2A2. | |
| In phosphate buffer (0.09% sodium azide) | |
| MGB1/MGC71974/UGB2 | |
| Rabbit | |
| 500 μL | |
| Primary | |
| This antibody reacts with mammaglobin from human breast tissue, no cross-reactivity with mammaglobin B, lipophilin and uteroglobin. | |
| Store at 4°C. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 31A5 | |
| Unconjugated | |
| Western Blot (1:50) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user. | |
| SCGB2A2 | |
| SCGB2A2 | |
| Recombinant protein corresponding to full length human SCGB2A2. | |
| RUO | |
| 4250 | |
| Human, Monkey, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction