Learn More
SCARF2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SCARF2.
Brand: Abnova PAB22849.100uL
Description
The protein encoded by this gene is similar to SCARF1/SREC-I, a scavenger receptor protein that mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). This protein has only little activity of internalizing modified low density lipoproteins (LDL), but it can interact with SCARF1 through its extracellular domain. The association of this protein with SCARF1 is suppressed by the presence of scavenger ligands. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Sequence: HHDLDNTLNCSFLEPPSGLEQPSPSWSSRASFSSFDTTDEGPVYCVPHEEAPAESRDPEVPTVPSpecifications
| SCARF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SCARF2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| NSR1/SREC-II/SREC2/SRECRP-1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 91179 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SCARF2 | |
| SCARF2 | |
| Recombinant protein corresponding to amino acids of human SCARF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only