missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARB1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Scarb1.
Brand: Abnova PAB12597.100uL
This item is not returnable.
View return policy
Specifications
| SCARB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Scarb1. | |
| In antiserum (0.05% sodium azide) | |
| AI120173/CD36/Cd36l1/Cla-1/Cla1/D5Ertd460e/SR-B1/SR-BI/SRBI/Srb1/mSR-BI | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to SR-BI. It can block human SR-BI/Cla-1 (tested in HepG2 cells). | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunoprecipitation (1:100) Blocking (1:500-1000) Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| Scarb1 | |
| Scarb1 | |
| Recombinant protein corresponding to Scarb1. | |
| RUO | |
| 20778 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction