Learn More
SCAP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SCAP.
Brand: Abnova PAB20276.100uL
Description
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis. [provided by RefSeq
Sequence: EVWDAIEGVLCCSSEEVSSGITALVFLDKRIVAARLNGSLDFFSLETHTALSPLQFRGTPGRGSSPASPVYSSSDTVACHLTHTVPCAHQKPITALKAAAGRLVTGSQDHTLRVFRLEDSpecifications
| SCAP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SCAP. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA0199 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 22937 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| SCAP | |
| SCAP | |
| Recombinant protein corresponding to amino acids of human SCAP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only