missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMSN1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SAMSN1.
Brand: Abnova PAB25682.100ug
This item is not returnable.
View return policy
Description
SAMSN1 is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains (Claudio et al., 2001 [PubMed 11536050]).[supplied by OMIM
Specifications
| SAMSN1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| SAMSN1 | |
| SAMSN1 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human SAMSN1. | |
| 100 μg | |
| Primary | |
| At least two isoforms of SAMSN are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SAMSN1. | |
| In PBS (0.05% sodium azide) | |
| HACS1/NASH1/SASH2/SH3D6B | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 64092 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction