missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SA-P, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial synthetic peptide of human SFTPA1.
Brand: Abnova PAB18301.100ug
This item is not returnable.
View return policy
Description
Sequence: KEQCVEMYTDGQWNDRSpecifications
| SA-P | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial synthetic peptide of human SFTPA1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| COLEC4/FLJ51913/SFTP1/SP-A/SP-A1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 653509 | |
| Human, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SFTPA1 | |
| SFTPA1 | |
| A synthetic peptide corresponding to amino acids 221-236 at of human SFTPA1. | |
| 100 μg | |
| Primary | |
| The synthetic peptide of this antibody is different from the rat sequence by two amino acids. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction