Learn More
S100A12 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant S100A12.
Brand: Abnova PAB28697.100uL
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities. [provided by RefSeq
Sequence: KLEEHLEGIVNIFHQYSVRKGHFDTLSKGELKQLLTKELANTIKNIKDKAVIDEIFQGLDANQDEQVDFQEFISLVAIALKAAHYHTHKESpecifications
| S100A12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant S100A12. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| CAAF1/CAGC/CGRP/ENRAGE/MRP6/p6 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6283 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/ml) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| S100A12 | |
| S100A12 | |
| Recombinant protein corresponding to amino acids of human S100A12. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only