Learn More
S100A12 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant S100A12.
Brand: Abnova PAB12047.100uL
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities. [provided by RefSeq
Specifications
| S100A12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant S100A12. | |
| In antiserum (0.09% sodium azide) | |
| CAAF1/CAGC/CGRP/ENRAGE/MRP6/p6 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| S100A12 | |
| S100A12 | |
| Recombinant protein corresponding to full length human S100A12. | |
| RUO | |
| 6283 | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only