missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1/S100B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant S100A1/S100B.
Brand: Abnova PAB11341.500ug
This item is not returnable.
View return policy
Specifications
| S100A1/S100B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant S100A1/S100B. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| S100A1 | |
| Recombinant protein corresponding to full length bovine S100A1/S100B. | |
| RUO | |
| 528735/525716 | |
| Bovine, Canine, Mouse, Rat | |
| Lyophilized |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:5000-1:25000) Western Blot (1:500-1:3000) Immunohistochemistry (1:200-1:2000) The optimal working dilution should be determined by the end user. | |
| MGC159706 | |
| Rabbit | |
| 500 μg | |
| Primary | |
| A 98% homology is noted for S100 alpha chain from primate sources. Mouse, rat and dog show 94% homology with the bovine S100 alpha sequence. Expect cross reactivity with S100 from most mammalian sources. Homologies for the S100 beta chain are similar. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction