Learn More
RRBP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RRBP1.
Brand: Abnova PAB20465.100uL
Description
Analysis of cDNA clones indicates that ribosome binding protein 1 may exist in different forms due to removal of tandem repeats, or partial intraexonic splicing of RRBP1. The form presented here is lacking the canine p180 ribosome-binding domain, NQGKKAEGAQ, which is tandemly repeated close to the N-terminus in other forms that haven't been fully characterized. RRBP1 has been excluded as a candidate gene in the cause of Alagille syndrome. Alternate splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: ELHSKLQSSEAEVRSKCEELSGLHGQLQEARAENSQLTERIRSIEALLEAGQARDAQDVQASQAEADQQQTRLKELESQVSGLEKEAIELREAVEQQKVKNNDLREKSpecifications
| RRBP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RRBP1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp586A1420/ES/130/ES130/FLJ36146/MGC157720/MGC157721/hES | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6238 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| RRBP1 | |
| RRBP1 | |
| Recombinant protein corresponding to amino acids of human RRBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only