Learn More
RPS8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RPS8.
Brand: Abnova PAB17576.100ug
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal tumors and colon polyps compared to matched normal colonic mucosa has been observed. This gene is co-transcribed with the small nucleolar RNA genes U38A, U38B, U39, and U40, which are located in its fourth, fifth, first, and second introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq
Specifications
| RPS8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RPS8. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| RPS8 | |
| A synthetic peptide corresponding to internal of human RPS8. | |
| RUO | |
| 6202 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| RPS8 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total RPS8 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only