Learn More
RPS6KB1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RPS6KB1.
Brand: Abnova PAB18199.100ug
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding this gene and overexpression of this kinase are seen in some breast cancer cell lines. Alternate translational start sites have been described and alternate transcriptional splice variants have been observed but have not been thoroughly characterized. [provided by RefSeq
Specifications
| RPS6KB1 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| RPS6KB1 | |
| RPS6KB1 | |
| A synthetic peptide corresponding to human RPS6KB1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to RPS6KB1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of RPS6KB1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PS6K/S6K/S6K1/STK14A/p70(S6K)-alpha/p70-S6K/p70-alpha | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 6198 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only