missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KA6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RPS6KA6.
Brand: Abnova PAB28694.100uL
This item is not returnable.
View return policy
Description
Sequence: PFKPASGKPDDTFCFDPEFTAKTPKDSPGLPASANAHQLFKGFSFVATSIAEEYKITPITSANVLPIVQINGNAAQFGEVYELKEDIGVGSYSVCKRCIHATTNMEFAVKIIDKSKSpecifications
| RPS6KA6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RPS6KA6. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| RSK4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27330 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| RPS6KA6 | |
| RPS6KA6 | |
| Recombinant protein corresponding to amino acids of human RPS6KA6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction