missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS27L, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RPS27L.
Brand: Abnova PAB17572.100ug
This item is not returnable.
View return policy
Description
This gene encodes a protein sharing 96% amino acid similarity with ribosomal protein S27, which suggests the encoded protein may be a component of the 40S ribosomal subunit. [provided by RefSeq
Specifications
| RPS27L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RPS27L. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| RPS27L | |
| A synthetic peptide corresponding to N-terminus of human RPS27L. | |
| RUO | |
| 51065 | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| RPS27L | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total RS27L protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction