Learn More
RPS15A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RPS15A.
Brand: Abnova PAB24528.100uL
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq
Sequence: TGRLNKCGVISPRFDVQLKDLESpecifications
| RPS15A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RPS15A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ27457/MGC111208/S15a | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6210 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| RPS15A | |
| RPS15A | |
| Recombinant protein corresponding to amino acids of human RPS15A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only