Learn More
RPGRIP1L, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RPGRIP1L.
Brand: Abnova PAB23540.100uL
Description
The protein encoded by this gene can localize to the basal body-centrosome complex or to primary cilia and centrosomes in ciliated cells. The encoded protein has been found to interact with nephrocystin-4. Defects in this gene are a cause of Joubert syndrome type 7 (JBTS7) and Meckel syndrome type 5 (MKS5). Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: KNTITLEVHQAYSTEYETIAACQLKFHEILEKSGRIFCTASLIGTKGDIPNFGTVEYWFRLRVPMDQAIRLYRERAKALGYITSNFKGPEHMQSLSQQAPKTAQLSSTDSTDGNLSpecifications
| RPGRIP1L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RPGRIP1L. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CORS3/DKFZp686C0668/JBTS7/KIAA1005/MKS5/NPHP8 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23322 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| RPGRIP1L | |
| RPGRIP1L | |
| Recombinant protein corresponding to amino acids of human RPGRIP1L. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only