Learn More
RP5-1000E104 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RP5-1000E10.4.
Brand: Abnova PAB21778.100uL
Description
SIKE interacts with IKK-epsilon (IKBKE; MIM 605048) and TBK1 (MIM 604834) and acts as a suppressor of TLR3 (MIM 603029) and virus-triggered interferon activation pathways (Huang et al., 2005 [PubMed 16281057]).[supplied by OMIM
Sequence: AEPVLKAHQSHSAEIESQIDRICEMGEVMRKAVQVDDDQFCKIQEKLAQLELENKELRELLSISSESLQARKENSMDSpecifications
| RP5-1000E104 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RP5-1000E10.4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp686A0768/FLJ21168/SIKE | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 80143 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| RP5-1000E10.4 | |
| RP5-1000E10.4 | |
| Recombinant protein corresponding to amino acids of human RP5-1000E10.4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only