Learn More
ROBLD3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ROBLD3.
Brand: Abnova PAB20265.100uL
Description
The product of this gene is highly conserved with a mouse protein associated with the cytoplasmic face of late endosomes and lysosomes. The mouse protein interacts with MAPK scaffold protein 1, a component of the mitogen-activated protein kinase pathway. In humans, a mutation in this gene has been associated with a primary immunodeficiency syndrome, and suggests a role for this protein in endosomal biogenesis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RPKALTQVLSQANTGGVQSTLLLNNEGSLLAYSGYGDTDARVTAAIASNIWAAYDRNGNQAFNEDNLKFILMDCMEGRVAITRVANLLLCMYAKETVGFGMLKAKAQALVQYLEEPSpecifications
| ROBLD3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ROBLD3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| ENDAP/HSPC003/MAPBPIP/MAPKSP1AP/p14 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 28956 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| ROBLD3 | |
| ROBLD3 | |
| Recombinant protein corresponding to amino acids of human ROBLD3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only