Learn More
RNF43, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RNF43.
Brand: Abnova PAB20409.100uL
Description
RNF43 is a HAP95 (AKAP8L; MIM 609475) binding ubiquitin ligase that promotes cell growth and is upregulated in colon cancer (Yagyu et al., 2004 [PubMed 15492824]; Sugiura et al., 2008 [PubMed 18313049]).[supplied by OMIM
Sequence: CVDPWLHQHRTCPLCMFNITEGDSFSQSLGPSRSYQEPGRRLHLIRQHPGHAHYHLPAAYLLGPSRSAVARPPRPGPFLPSQEPGMGPRHHRFPRAAHPRAPGEQQRLAGAQHPYAQGWGLSHLQSTSQHSpecifications
| RNF43 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RNF43. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp781H02126/DKFZp781H0392/FLJ20315/FLJ77466/FLJ99338/MGC125630/RNF124/URCC | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54894 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| RNF43 | |
| RNF43 | |
| Recombinant protein corresponding to amino acids of human RNF43. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only