Learn More
RNF38, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RNF38.
Brand: Abnova PAB20851.100uL
Description
This gene encodes a protein with a coiled-coil motif and a RING-H2 motif (C3H2C2) at its carboxy-terminus. The RING motif is a zinc-binding domain found in a large set of proteins playing roles in diverse cellular processes including oncogenesis, development, signal transduction, and apoptosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: VVFSGQHLPVCSVPPPMLQACSVQHLPVPYAAFPPLISSDPFLIHPPHLSPHHPPHLPPPGQFVPFQTQQSRSPLQRIENEVELLGEHLPVGGFTYPPSAHPPTLPPSAPLSpecifications
| RNF38 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RNF38. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ21343 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 152006 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| RNF38 | |
| RNF38 | |
| Recombinant protein corresponding to amino acids of human RNF38. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only