Learn More
RNASEL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RNASEL.
Brand: Abnova PAB28675.100uL
Description
This gene encodes a component of the interferon-regulated 2-5A system that functions in the antiviral and antiproliferative roles of interferons. Mutations in this gene have been associated with predisposition to prostate cancer and this gene is a candidate for the hereditary prostate cancer 1 (HPC1) allele. [provided by RefSeq
Sequence: HGAKEDFHPPAEDWKPQSSHWGAALKDLHRIYRPMIGKLKFFIDEKYKIADTSEGGIYLGFYEKQEVAVKTFCEGSPRAQREVSCLQSSRENSHLVTFYGSESHRGHLFVCVTLCEQTLEACLDVHRGEDVENEEDEFARNVLSSISpecifications
| RNASEL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RNASEL. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| DKFZp781D08126/MGC104972/MGC133329/PRCA1/RNS4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6041 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry Western Blot The optimal working dilution should be determined by the end user. | |
| RNASEL | |
| RNASEL | |
| Recombinant protein corresponding to amino acids of human RNASEL. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only