missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Ripk3.
Brand: Abnova PAB12850.100ug
This item is not returnable.
View return policy
Description
Sequence: AQFGRGRGWQPFHKSpecifications
| RIPK3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Ripk3. | |
| In PBS (0.02% sodium azide) | |
| 2610528K09Rik/AW107945/Rip3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| An approximately 57 KDa band can be detected. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Ripk3 | |
| Ripk3 | |
| A synthetic peptide corresponding to amino acids 473-486 of mouse Ripk3. | |
| RUO | |
| 56532 | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction