Learn More
RGR Rabbit anti-Horse, Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RGR.
Brand: Abnova PAB26528.50ug
Description
This gene encodes a putative retinal G-protein coupled receptor. The gene is a member of the opsin subfamily of the 7 transmembrane, G-protein coupled receptor 1 family. Like other opsins which bind retinaldehyde, it contains a conserved lysine residue in the seventh transmembrane domain. The protein acts as a photoisomerase to catalyze the conversion of all-trans-retinal to 11-cis-retinal. The reverse isomerization occurs with rhodopsin in retinal photoreceptor cells. The protein is exclusively expressed in tissue adjacent to retinal photoreceptor cells, the retinal pigment epithelium and Mueller cells. This gene may be associated with autosomal recessive and autosomal dominant retinitis pigmentosa (arRP and adRP, respectively). Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| RGR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RGR. | |
| In PBS (0.09% sodium azide) | |
| RGR | |
| A synthetic peptide corresponding to 16 amino acids at 2nd extracellular domain of human RGR. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (9-17 ug/mL) The optimal working dilution should be determined by the end user. | |
| RGR | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 5995 | |
| Equine, Human, Monkey | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only