Learn More
RFX5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RFX5.
Brand: Abnova PAB11300.100ug
Description
A lack of MHC-II expression results in a severe immunodeficiency syndrome called MHC-II deficiency, or the bare lymphocyte syndrome (BLS; MIM 209920). At least 4 complementation groups have been identified in B-cell lines established from patients with BLS. The molecular defects in complementation groups B, C, and D all lead to a deficiency in RFX, a nuclear protein complex that binds to the X box of MHC-II promoters. The lack of RFX binding activity in complementation group C results from mutations in the RFX5 gene encoding the 75kDa subunit of RFX (Steimle et al., 1995). RFX5 is the fifth member of the growing family of DNA-binding proteins sharing a novel and highly characteristic DNA-binding domain called the RFX motif. Multiple alternatively spliced transcript variants have been found but the full-length natures of only two have been determined. [provided by RefSeq
Specifications
| RFX5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RFX5. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| RFX5 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 320-494 of human RFX5. | |
| RUO | |
| 5993 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Gel Shift, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Gel Supershift assay (0.5-1.0 ul per assay) The optimal working dilution should be determined by the end user. | |
| RFX5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only