Learn More
RFK, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RFK.
Brand: Abnova PAB2744.400uL
Description
Riboflavin kinase (RFK) is an essential enzyme catalyzing the phosphorylation of riboflavin (vitamin B2) to form flavin mononucleotide (FMN), an obligatory step in vitamin B2 utilization and flavin cofactor synthesis (Karthikeyan et al., 2003 [PubMed 12623014]).[supplied by OMIM
Specifications
| RFK | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RFK. | |
| In PBS (0.09% sodium azide) | |
| FLJ11149/RIFK | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 55312 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| RFK | |
| RFK | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human RFK. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only