missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFA1, RFA2, RFA3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RFA1, RFA2, RFA3.
Brand: Abnova PAB13584.50uL
This item is not returnable.
View return policy
Specifications
| RFA1, RFA2, RFA3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RFA1, RFA2, RFA3. | |
| Lyophilized from serum | |
| Recombinant protein corresponding to Saccharomyces cerevisiae RFA1/RFA2/RFA3. | |
| RUO | |
| Yeast | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:20000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Store at -20°C on dry atmosphere. After reconstitution with 100μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction