Learn More
RC3H1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RC3H1.
Brand: Abnova PAB27394.100ug
Description
RC3H1, or roquin, encodes a highly conserved member of the RING type ubiquitin ligase protein family (Vinuesa et al., 2005 [PubMed 15917799]). The roquin protein is distinguished by the presence of a CCCH zinc finger found in RNA-binding proteins, and localization to cytosolic RNA granules implicated in regulating mRNA translation and stability.[supplied by OMIM
Specifications
| RC3H1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| RC3H1 | |
| RC3H1 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human RC3H1. | |
| 100 μg | |
| Primary | |
| At least three isoforms of RC3H1 are known to exist; this antibody will detect all three isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of RC3H1. | |
| In PBS (0.02% sodium azide) | |
| KIAA2025/RNF198/ROQUIN | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 149041 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only