Learn More
RBM22, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RBM22
Brand: Abnova PAB28467.100uL
Description
This gene encodes an RNA binding protein. The encoded protein may play a role in cell division and may be involved in pre-mRNA splicing. Related pseudogenes exist on chromosomes 6, 7, 9, 13, 16, 18, and X. [provided by RefSeq
Sequence: NTYNRQNWEDADFPILCQTCLGENPYIRMTKEKYGKECKICARPFTVFRWCPGVRMRFKKTEVCQTCSKLKNVCQTCLLDLEYGLPIQVRDAGLSFKDDMPKSDVNKEYYTQNMEREISNSDGTRPVGMLGKATSTSDMLLKLSpecifications
| RBM22 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RBM22 | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FLJ10290/ZC3H16/fSAP47 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55696 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:20-1:50) Western Blot(1:100-1:250) Immunofluorescence(1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| RBM22 | |
| RBM22 | |
| Recombinant protein corresponding to amino acids of human RBM22 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only