Learn More
RBM15B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RBM15B.
Brand: Abnova PAB23043.100uL
Description
Members of the SPEN (Split-end) family of proteins, including RBM15B, have repressor function in several signaling pathways and may bind to RNA through interaction with spliceosome components (Hiriart et al., 2005 [PubMed 16129689]).[supplied by OMIM
Sequence: GKKARDSERNHRTTEAEPKPLEEPKHETKKLKNLSEYAQTLQLGWNGLLVLKNSCFPTSMHILEGDQGVISSLLKDHTSGSpecifications
| RBM15B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant RBM15B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| HUMAGCGB/OTT3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 29890 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| RBM15B | |
| RBM15B | |
| Recombinant protein corresponding to amino acids of human RBM15B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only