Learn More
RBKS, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RBKS.
Brand: Abnova PAB2742.400uL
Description
The ribokinase encoded by this gene belongs to the pfkB family of carbohydrate kinases. It phosphorylates ribose to form ribose-5-phosphate in the presence of ATP and magnesium as a first step in ribose metabolism. [provided by RefSeq
Specifications
| RBKS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of RBKS. | |
| In PBS (0.09% sodium azide) | |
| DKFZp686G13268/RBSK | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 64080 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| RBKS | |
| RBKS | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human RBKS. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only