missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgM Heavy Chain Secondary Antibody, Texas Red, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73710
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| Texas Red | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| IGHM | |
| Rat IgM u chain | |
| 1.5 mg | |
| Secondary | |
| No reaction was observed against other rat heavy or light chain proteins. | |
| Rat | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction