missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgG2b Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69611
This item is not returnable.
View return policy
Description
IgG2b Polyclonal antibody specifically detects IgG2b in Rat samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
Specifications
| IgG2b | |
| Polyclonal | |
| Unconjugated | |
| Whole antiserum | |
| Pools of purified homogenous IgG2b isolated from rat serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunoprecipitation, Immunoelectrophoresis | |
| LYOPH | |
| ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| Secondary | |
| Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction