Learn More
Rabbit anti-Rat IgG whole molecule, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB10873.2mg
Product Code. 16283160
  • £218.00 / 2mg
Specifications
Rabbit anti-Rat IgG whole molecule
Polyclonal
Rabbit anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red.
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free))
Rat IgG whole molecule
2 mg
Rat
Lyophilized
Flow Cytometry, Immunofluorescence
Texas Red
The optimal working dilution should be determined by the end user.
Rabbit
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.