Learn More
Rabbit anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75419-0.5mg
 View more versions of this product
Product Code. 18576671
  • £121.00 / 0.50mg
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Unconjugated
Purified Rat IgG (H&L)
0.5 mg
Adaptive Immunity, Immunology
Based on IEP, this antibody reacts with heavy gamma chains on rat IgG and light chains on all rat immunoglobulins. This antibody has been pre-adsorbed against human serum proteins.
Store at 4°C. Do not freeze.
Immunoassay
4.5 mg/mL
Rabbit
Affinity purified
RUO
Secondary
Rat